Learn More
Selleck Chemical LLC Aminopterin-E1003-5MG
SDP
Catalog No. 503135348
Encompass_Preferred
Aminopterin (4-amino Folic Acid 4-amino PGA 4-Aminofolic acid 4-Aminopteroylglutamic acid Aminopteroylglutamic acid APGA NSC 739) a folic acid antagonist is a competitive inhibitor of dihydrofolate reductase (DHFR) with Ki of 3 7 pM
Catalog No. 50-313-5348 Supplier Selleck Chemical LLC Supplier No. E10035MG
Aminopterin (4-amino Folic Acid 4-amino PGA 4-Aminofolic acid 4-Aminopteroylglutamic acid Aminopteroylglutamic acid APGA NSC 739) a folic acid antagonist is a competitive inhibitor of dihydrofolate reductase (DHFR) with Ki of 3 7 pM