Learn More
Enzo Life Sciences Aminopurvalanol A (1mg). CAS: 220792-57-4
SDP
Supplier:  Enzo Life Sciences ALX270249M001
Encompass_Preferred
Cell permeable, reversible and competitive inhibitor of CDK1/cyclin B (IC50=33nM), CDK2/cyclin A (IC50=33nM), CDK2/cyclin E (IC50=28nM) and CDK5/p35 (IC50=20nM). Alternative name: (2R)-2-((6-((3-Amino-5-chlorophenyl)amino)-9-(1-methylethyl)-9H-purin-2-yl)amino)-3-methyl-1-butanol, NG-97. Formula: C19H26N7OCl. MW: 403.9. Purity: ≥97% (HPLC). Appearance: White to brownish solid. Solubility: Soluble in DMSO or methanol. Long Term Storage: +4°C. Handling: Protect from light. Packaged under inert gas.
Catalog No. 50-200-8445