Learn More
Medchemexpress LLC Aminopurvalanol A | 220792-57-4 | MFCD06798384 | 98.0% | 403.91 g/mol | C19H26ClN7O | 1 ML
SDP
Catalog No. 5000241845
Encompass_Preferred
Aminopurvalanol A is a selective, cell-permeable cyclin-dependent kinase (CDK) inhibitor used in research to probe cell cycle control and apoptosis pathways. It is supplied as solid material in multiple milligram scales and as a ready-to-use 10 mM solution in DMSO, with documented purity and storage recommendations for experimental stability.
  • Selective inhibitor of cyclin-dependent kinases for mechanistic studies.
  • Cell-permeable small molecule suitable for cellular assays.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • High reported purity to support reproducible results.
  • Defined storage conditions to maintain compound stability.
Catalog No. 50-002-41845 Supplier Medchemexpress LLC Supplier No. HY10401310MM 1ML
Aminopurvalanol A is a selective, cell-permeable cyclin-dependent kinase (CDK) inhibitor used in research to probe cell cycle control and apoptosis pathways. It is supplied as solid material in multiple milligram scales and as a ready-to-use 10 mM solution in DMSO, with documented purity and storage recommendations for experimental stability.
  • Selective inhibitor of cyclin-dependent kinases for mechanistic studies.
  • Cell-permeable small molecule suitable for cellular assays.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • High reported purity to support reproducible results.
  • Defined storage conditions to maintain compound stability.