Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aminothiazole | 96-50-4 | MFCD00005325 | 99.8% | 100.14 g/mol | C3H4N2S | 1KG
SDP

Catalog No. 5000238870
Encompass_Preferred

Aminothiazole (2-aminothiazole) is a heterocyclic amine used as a building block in medicinal chemistry and as a reagent in biochemical research. Supplied as a high-purity solid for research use only, it is employed in the synthesis of biologically active molecules and in studies involving bacterial, fungal, and viral proteases.

  • High purity: 99.8%.
  • Molecular weight: 100.14 g/mol.
  • Chemical formula: C3H4N2S.
  • Available in multiple pack sizes, including 1 kg solid.
  • Supplied with supporting documents such as datasheet and SDS.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-38870 Supplier Medchemexpress LLC Supplier No. HY123961KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminothiazole (2-aminothiazole) is a heterocyclic amine used as a building block in medicinal chemistry and as a reagent in biochemical research. Supplied as a high-purity solid for research use only, it is employed in the synthesis of biologically active molecules and in studies involving bacterial, fungal, and viral proteases.

  • High purity: 99.8%.
  • Molecular weight: 100.14 g/mol.
  • Chemical formula: C3H4N2S.
  • Available in multiple pack sizes, including 1 kg solid.
  • Supplied with supporting documents such as datasheet and SDS.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.