Learn More
Novus Biologicals™ AminTRAP HIS Prepacked Columns

Description
- Packed with Ni-NTA resin
- AminTRAP HIS Prepacked Columns have a standard interface and can be adapted to chromatography systems such as AKTA
- This product is for research use only and is not approved for use in humans or in clinical diagnosis
Specifications
Specifications
| Formulation | Packed with Ni-NTA resin in vials |
| For Use With (Application) | Affinity purification of HIS-tagged proteins. Can be adapted to chromatography systems such as AKTA. |
| Content And Storage | 4°C. Do not freeze. |
| Quantity | 5 x 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.