Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amiodarone-d10 hydrochloride | 1261393-77-4 | MFCD25976575 | 98.8% | 691.83 | C25H20D10ClI2NO3 | 5 MG
SDP

Catalog No. 5000220991
Encompass_Preferred

Amiodarone-d10 hydrochloride is a deuterium-labeled analogue of the antiarrhythmic compound amiodarone, provided as the hydrochloride salt and intended for research use as a stable isotope reference standard and analytical internal standard for LC-MS applications.

  • Deuterium-labeled (d10) for use as a stable isotope internal standard.
  • Hydrochloride salt supplied as a solid for convenient handling and storage.
  • High purity: 98.8% by analysis, suitable for quantitative assays.
  • Molecular weight ~691.83 Da, molecular formula C25H20D10ClI2NO3.
  • Available in small research-scale quantities for analytical method development.

Catalog No. 50-002-20991 Supplier Medchemexpress LLC Supplier No. HY14187S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amiodarone-d10 hydrochloride is a deuterium-labeled analogue of the antiarrhythmic compound amiodarone, provided as the hydrochloride salt and intended for research use as a stable isotope reference standard and analytical internal standard for LC-MS applications.

  • Deuterium-labeled (d10) for use as a stable isotope internal standard.
  • Hydrochloride salt supplied as a solid for convenient handling and storage.
  • High purity: 98.8% by analysis, suitable for quantitative assays.
  • Molecular weight ~691.83 Da, molecular formula C25H20D10ClI2NO3.
  • Available in small research-scale quantities for analytical method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.