Learn More
Medchemexpress LLC Amiodarone-d10 hydrochloride | 1261393-77-4 | MFCD34567863 | 98.8% | C25H20D10ClI2NO3 | 10MG
SDP
Catalog No. 5000261294
Encompass_Preferred
Amiodarone-d10 hydrochloride is a deuterium-labeled analog of the antiarrhythmic agent amiodarone, supplied as the hydrochloride salt. It is used as a stable-isotope internal standard or tracer in analytical chemistry, pharmacokinetic studies, and other research applications.
  • Deuterium-labeled internal standard for quantitative analysis
  • Hydrochloride salt supplied as a solid
  • High purity (98.8%) for reliable analytical performance
  • Suitable for pharmacokinetic and tracer studies
  • Available in laboratory-scale packaging for research use
Catalog No. 50-002-61294 Supplier Medchemexpress LLC Supplier No. HY14187S10MG
Amiodarone-d10 hydrochloride is a deuterium-labeled analog of the antiarrhythmic agent amiodarone, supplied as the hydrochloride salt. It is used as a stable-isotope internal standard or tracer in analytical chemistry, pharmacokinetic studies, and other research applications.
  • Deuterium-labeled internal standard for quantitative analysis
  • Hydrochloride salt supplied as a solid
  • High purity (98.8%) for reliable analytical performance
  • Suitable for pharmacokinetic and tracer studies
  • Available in laboratory-scale packaging for research use