Learn More
Medchemexpress LLC Amiodarone-d4 hydrochloride | 1216715-80-8 | MFCD08064189 | 99.6% | 685.80 | C25H26D4ClI2NO3 | 5 MG
SDP
Catalog No. 5000221700
Encompass_Preferred
Deuterium-labeled amiodarone hydrochloride provided as a solid isotopically labeled standard for analytical and pharmacological research. It is used as an internal standard or tracer in LC-MS/MS, pharmacokinetic, and metabolic studies where a stable isotope analogue of amiodarone is required.
  • Stable isotope-labeled internal standard for quantitative analysis.
  • High purity, 99.6%.
  • Solid form for convenient storage and accurate weighing.
  • Soluble in DMSO, DMF, and ethanol.
  • Suitable for LC-MS/MS, pharmacokinetic, and tracer studies.
Catalog No. 50-002-21700 Supplier Medchemexpress LLC Supplier No. HY14188S5MG
Deuterium-labeled amiodarone hydrochloride provided as a solid isotopically labeled standard for analytical and pharmacological research. It is used as an internal standard or tracer in LC-MS/MS, pharmacokinetic, and metabolic studies where a stable isotope analogue of amiodarone is required.
  • Stable isotope-labeled internal standard for quantitative analysis.
  • High purity, 99.6%.
  • Solid form for convenient storage and accurate weighing.
  • Soluble in DMSO, DMF, and ethanol.
  • Suitable for LC-MS/MS, pharmacokinetic, and tracer studies.