Learn More
Medchemexpress LLC Amiodarone (hydrochloride) | 19774-82-4 | 99.9% | C25H30ClI2NO3 | 1 ML
SDP
Catalog No. 5000353648
Encompass_Preferred
Amiodarone hydrochloride is a potent antiarrhythmic agent primarily used for severe cardiac arrhythmias. It belongs to Class III antiarrhythmics, characterized by its ability to prolong the action potential duration and refractory period in myocardial tissues. This product is provided as a 10 mM solution in DMSO, offering convenience for various research applications.
  • Potent antiarrhythmic agent
  • Prolongs action potential and refractory period
  • Supplied as a 10 mM solution in DMSO
  • Suitable for in vitro and in vivo research
  • Recommended storage at -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light) when in solvent
Catalog No. 50-003-53648 Supplier Medchemexpress LLC Supplier No. HY1418810MM/1ML
Amiodarone hydrochloride is a potent antiarrhythmic agent primarily used for severe cardiac arrhythmias. It belongs to Class III antiarrhythmics, characterized by its ability to prolong the action potential duration and refractory period in myocardial tissues. This product is provided as a 10 mM solution in DMSO, offering convenience for various research applications.
  • Potent antiarrhythmic agent
  • Prolongs action potential and refractory period
  • Supplied as a 10 mM solution in DMSO
  • Suitable for in vitro and in vivo research
  • Recommended storage at -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light) when in solvent