Learn More
Medchemexpress LLC Amiodarone hydrochloride | 19774-82-4 | C25H30ClI2NO3 | 500 MG
SDP
Catalog No. 5000354575
Encompass_Preferred
Amiodarone hydrochloride is a benzofuran-based Class III antiarrhythmic agent. It has been shown to inhibit WT outward IhERG tails and induce cell proliferation and myofibroblast differentiation via ERK1/2 and p38 MAPK signaling in fibroblasts. It is suitable for research into both supraventricular and ventricular arrhythmias.
  • Benzofuran-based Class III antiarrhythmic agent
  • Inhibits WT outward IhERG tails with an IC50 of approximately 45 nM
  • Induces cell proliferation and myofibroblast differentiation via ERK1/2 and p38 MAPK signaling
  • Useful in research for supraventricular and ventricular arrhythmias
Catalog No. 50-003-54575 Supplier Medchemexpress LLC Supplier No. HY14188500MG
Amiodarone hydrochloride is a benzofuran-based Class III antiarrhythmic agent. It has been shown to inhibit WT outward IhERG tails and induce cell proliferation and myofibroblast differentiation via ERK1/2 and p38 MAPK signaling in fibroblasts. It is suitable for research into both supraventricular and ventricular arrhythmias.
  • Benzofuran-based Class III antiarrhythmic agent
  • Inhibits WT outward IhERG tails with an IC50 of approximately 45 nM
  • Induces cell proliferation and myofibroblast differentiation via ERK1/2 and p38 MAPK signaling
  • Useful in research for supraventricular and ventricular arrhythmias