Learn More
Medchemexpress LLC Amiodarone hydrochloride (Standard) | 19774-82-4 | C25H30CII2NO3 | 25 MG
SDP
Catalog No. 5000352540
Encompass_Preferred
Amiodarone hydrochloride (Standard) is an analytical standard intended for research and analytical applications. This benzofuran-based Class III antiarrhythmic agent inhibits WT outward IhERG tails with an IC50 of approximately 45 nM. It also induces cell proliferation and myofibroblast differentiation through ERK1/2 and p38 MAPK signaling in fibroblasts. Amiodarone hydrochloride is utilized in the research of both supraventricular and ventricular arrhythmias. Please store the product under the recommended conditions found in the Certificate of Analysis.
  • Analytical standard
  • Intended for research and analytical applications
  • Benzofuran-based Class III antiarrhythmic agent
  • Inhibits WT outward IhERG tails
  • Induces cell proliferation and myofibroblast differentiation via ERK1/2 and p38 MAPK signaling in fibroblasts
  • Used in research of supraventricular and ventricular arrhythmias
Catalog No. 50-003-52540 Supplier Medchemexpress LLC Supplier No. 25MGHY14188R
Amiodarone hydrochloride (Standard) is an analytical standard intended for research and analytical applications. This benzofuran-based Class III antiarrhythmic agent inhibits WT outward IhERG tails with an IC50 of approximately 45 nM. It also induces cell proliferation and myofibroblast differentiation through ERK1/2 and p38 MAPK signaling in fibroblasts. Amiodarone hydrochloride is utilized in the research of both supraventricular and ventricular arrhythmias. Please store the product under the recommended conditions found in the Certificate of Analysis.
  • Analytical standard
  • Intended for research and analytical applications
  • Benzofuran-based Class III antiarrhythmic agent
  • Inhibits WT outward IhERG tails
  • Induces cell proliferation and myofibroblast differentiation via ERK1/2 and p38 MAPK signaling in fibroblasts
  • Used in research of supraventricular and ventricular arrhythmias