Learn More
Cayman Chemical Amiodarone-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 1216715-80-8, 1 mg
SDP
Catalog No. 502891358
Encompass_Preferred
An internal standard for the quantification of amiodarone by GC- or LC-MS
Catalog No. 50-289-1358 Supplier Cayman Chemical Supplier No. 100106681 MG
An internal standard for the quantification of amiodarone by GC- or LC-MS