Learn More
Cayman Chemical Amiodaroned4 hydrochloride
SDP
Catalog No. 502888494
Encompass_Preferred
An internal standard for the quantification of amiodarone by GC- or LC-MS
Catalog No. 50-288-8494 Supplier Cayman Chemical Supplier No. 100106685 MG
An internal standard for the quantification of amiodarone by GC- or LC-MS