Learn More
Medchemexpress LLC Amiprofos methyl (BAY-NTN 6867) | 36001-88-4 | 99.4% | 304.30 | 25 MG
SDP
Catalog No. 5000439727
Encompass_Preferred
Amiprofos methyl is a phosphoric amide herbicide. It is a specific and potent antimicrotubule agent that directly poisons microtubule dynamics in plant cells.
  • Phosphoric amide herbicide
  • Specific and potent antimicrotubule agent
  • Directly poisons microtubule dynamics in plant cells
Catalog No. 50-004-39727 Supplier Medchemexpress LLC Supplier No. 25MGHY111939
Amiprofos methyl is a phosphoric amide herbicide. It is a specific and potent antimicrotubule agent that directly poisons microtubule dynamics in plant cells.
  • Phosphoric amide herbicide
  • Specific and potent antimicrotubule agent
  • Directly poisons microtubule dynamics in plant cells