Learn More
AdipoGen Amisulbrom
SDP
Catalog No. 502561775
Encompass_Preferred
Chemical. CAS 348635-87-0. Formula C13H13BrFN5O4S2. MW 466.31. Synthetic. Oomycete-specific sulphonamide fungicide which acts by inhibiting mitochondrial complex III activity Complex III cytochrome bc1 ubiquinone reductase through binding to the Qi centre and impairing respiration and energy production. Categorized as quinine insider inhibitor QII. Shown to have anti-fouling activity. Compound can be used as analytical reference material.
Catalog No. 50-256-1775 Supplier AdipoGen Supplier No. CDXA0262M100
Chemical. CAS 348635-87-0. Formula C13H13BrFN5O4S2. MW 466.31. Synthetic. Oomycete-specific sulphonamide fungicide which acts by inhibiting mitochondrial complex III activity Complex III cytochrome bc1 ubiquinone reductase through binding to the Qi centre and impairing respiration and energy production. Categorized as quinine insider inhibitor QII. Shown to have anti-fouling activity. Compound can be used as analytical reference material.