Learn More
FUJIFILM Biosciences Amisulbrom Standard
Catalog No. 5000450361
Encompass_Preferred
Reagent for Pesticide Residue Analysis Amisulbrom is a novel sulfamoyl-triazole fungicide which has been found to be effective against oomycota and powdery mildew in relatively small amounts Amisulbrom affects the Qi site of mitochondrial complex III of the electron transport chain of oomycota This is believed to be the reason why amisulbrom is effective against fungi that have developed a resistance against conventional phenylamide and strobilurin fungicides White crystalline powder or powder Amisulbrom Standard with 98 0% purity suitable for HPLC and qNMR methods in pesticide residue analysis Solid form shipped and stored under cool inert and light-protected conditions
Catalog No. 50-004-50361 Supplier FUJIFILM Biosciences Supplier No. 01624761
Reagent for Pesticide Residue Analysis Amisulbrom is a novel sulfamoyl-triazole fungicide which has been found to be effective against oomycota and powdery mildew in relatively small amounts Amisulbrom affects the Qi site of mitochondrial complex III of the electron transport chain of oomycota This is believed to be the reason why amisulbrom is effective against fungi that have developed a resistance against conventional phenylamide and strobilurin fungicides White crystalline powder or powder Amisulbrom Standard with 98 0% purity suitable for HPLC and qNMR methods in pesticide residue analysis Solid form shipped and stored under cool inert and light-protected conditions