Learn More
Medchemexpress LLC Amisulpride | 71675-85-9 | MFCD00866691 | 99.8% | C17H27N3O4S | 200 MG
SDP
Catalog No. 5000353810
Encompass_Preferred
Amisulpride is a racemic (50:50) mixture of R-Amisulpride and S-Amisulpride. It acts as a dopamine D2/D3 receptor antagonist with Ki values of 2.8 nM for human dopamine D2 and 3.2 nM for human dopamine D3 receptors. Amisulpride also functions as a 5-HT7 receptor antagonist with a Ki of 44 nM, making it suitable for use in psychiatric research.
  • Racemic mixture
  • Dopamine D2/D3 receptor antagonist
  • 5-HT7 receptor antagonist
  • Suitable for psychiatric research
  • D3 receptor Ki: 3.2 nM
  • Human D2 receptor Ki: 2.8 nM
  • 5-HT7 receptor Ki: 44 nM
  • Inhibits quinpirole-elicited [3H]thymidine incorporation
  • Increases dopamine release from rat striatum slices
  • Increases dopamine synthesis in rat striatum and limbic system
  • Increases extracellular dopamine levels
  • Decreases striatal ACh levels
  • Increases duration of swimming behavior
Catalog No. 50-003-53810 Supplier Medchemexpress LLC Supplier No. HY14545200MG
Amisulpride is a racemic (50:50) mixture of R-Amisulpride and S-Amisulpride. It acts as a dopamine D2/D3 receptor antagonist with Ki values of 2.8 nM for human dopamine D2 and 3.2 nM for human dopamine D3 receptors. Amisulpride also functions as a 5-HT7 receptor antagonist with a Ki of 44 nM, making it suitable for use in psychiatric research.
  • Racemic mixture
  • Dopamine D2/D3 receptor antagonist
  • 5-HT7 receptor antagonist
  • Suitable for psychiatric research
  • D3 receptor Ki: 3.2 nM
  • Human D2 receptor Ki: 2.8 nM
  • 5-HT7 receptor Ki: 44 nM
  • Inhibits quinpirole-elicited [3H]thymidine incorporation
  • Increases dopamine release from rat striatum slices
  • Increases dopamine synthesis in rat striatum and limbic system
  • Increases extracellular dopamine levels
  • Decreases striatal ACh levels
  • Increases duration of swimming behavior