Learn More
Medchemexpress LLC Amisulpride-d5 | 1216626-17-3 | 98.27% | C17H22D5N3O4S | 1 MG
SDP
Catalog No. 5000356427
Encompass_Preferred
Amisulpride-d5 is the deuterium-labeled form of Amisulpride, a dopamine D2/D3 receptor antagonist. It has Ki values of 2.8 nM for human dopamine D2 and 3.2 nM for human dopamine D3. This product is suitable for research applications, including use as a tracer and an internal standard for quantitative analysis.
  • Deuterium labeled
  • Dopamine D2/D3 receptor antagonist
  • High purity: 98.27%
  • Appearance: solid, white to off-white
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Valuable for pharmacokinetic and metabolic studies
Catalog No. 50-003-56427 Supplier Medchemexpress LLC Supplier No. HY14545S1MG
Amisulpride-d5 is the deuterium-labeled form of Amisulpride, a dopamine D2/D3 receptor antagonist. It has Ki values of 2.8 nM for human dopamine D2 and 3.2 nM for human dopamine D3. This product is suitable for research applications, including use as a tracer and an internal standard for quantitative analysis.
  • Deuterium labeled
  • Dopamine D2/D3 receptor antagonist
  • High purity: 98.27%
  • Appearance: solid, white to off-white
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Valuable for pharmacokinetic and metabolic studies