Learn More
Medchemexpress LLC Amisulpride-d5 | 1216626-17-3 | 98.8% | 374.51 | C17H22D5N3O4S | 5 MG
SDP
Catalog No. 5000221105
Encompass_Preferred
Amisulpride-d5 is the deuterium-labeled form of amisulpride intended for research use as a tracer or internal standard. It maintains the pharmacological profile as a dopamine D2/D3 receptor antagonist and is provided as a white to off-white solid suitable for quantitative analyses by NMR, GC-MS, or LC-MS.
  • Deuterium-labeled for isotope-dilution assays.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • High purity (≈98.8%) appropriate for analytical applications.
  • Soluble in DMSO and sparingly soluble in water; ultrasonic assistance recommended.
  • Stable as a powder at -20°C for long-term storage.
Catalog No. 50-002-21105 Supplier Medchemexpress LLC Supplier No. HY14545S5MG
Amisulpride-d5 is the deuterium-labeled form of amisulpride intended for research use as a tracer or internal standard. It maintains the pharmacological profile as a dopamine D2/D3 receptor antagonist and is provided as a white to off-white solid suitable for quantitative analyses by NMR, GC-MS, or LC-MS.
  • Deuterium-labeled for isotope-dilution assays.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • High purity (≈98.8%) appropriate for analytical applications.
  • Soluble in DMSO and sparingly soluble in water; ultrasonic assistance recommended.
  • Stable as a powder at -20°C for long-term storage.