Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amisulpride-d5 | 1216626-17-3 | 98.8% | C17H22D5N3O4S | 10MG
SDP

Catalog No. 5000261866
Encompass_Preferred

Amisulpride-d5 is the deuterium-labeled analogue of amisulpride used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. Supplied as a high-purity deuterated solid, it is intended for analytical and research applications where accurate quantitation is required.

  • Deuterium-labeled internal standard for mass spectrometry and NMR.
  • High deuteration purity (≈98.8%) for reliable quantitation.
  • Suitable as tracer and internal standard in bioanalytical assays.
  • Supplied as a 10 mg solid for small-scale analyses.
  • Molecular formula C17H22D5N3O4S; molecular weight 374.51 g/mol.

Catalog No. 50-002-61866 Supplier Medchemexpress LLC Supplier No. HY14545S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amisulpride-d5 is the deuterium-labeled analogue of amisulpride used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. Supplied as a high-purity deuterated solid, it is intended for analytical and research applications where accurate quantitation is required.

  • Deuterium-labeled internal standard for mass spectrometry and NMR.
  • High deuteration purity (≈98.8%) for reliable quantitation.
  • Suitable as tracer and internal standard in bioanalytical assays.
  • Supplied as a 10 mg solid for small-scale analyses.
  • Molecular formula C17H22D5N3O4S; molecular weight 374.51 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.