Learn More
Medchemexpress LLC Amisulpride (hydrochloride) | 81342-13-4 | 99.8% | C17H28ClN3O4S | 25 MG
SDP
Catalog No. 5000354610
Encompass_Preferred
Amisulpride hydrochloride is a dopamine D2/D3 receptor antagonist, with Ki values of 2.8 nM for human dopamine D2 and 3.2 nM for human dopamine D3. It is an atypical antagonist intended for research use only.
  • Inhibits quinpirole-elicited [3H]thymidine incorporation with an IC50 value of 22±3 nM.
  • Slightly increases [3H]dopamine release from rat striatum slices.
  • Reduces dopamine levels in the striatum or limbic system at high doses.
  • Increases dopamine synthesis in rat striatum and limbic system.
  • Increases extracellular dopamine levels.
  • Decreases striatal ACh levels.
  • Increases the duration of swimming behavior in acute studies.
Catalog No. 50-003-54610 Supplier Medchemexpress LLC Supplier No. 25MGHY14545A
Amisulpride hydrochloride is a dopamine D2/D3 receptor antagonist, with Ki values of 2.8 nM for human dopamine D2 and 3.2 nM for human dopamine D3. It is an atypical antagonist intended for research use only.
  • Inhibits quinpirole-elicited [3H]thymidine incorporation with an IC50 value of 22±3 nM.
  • Slightly increases [3H]dopamine release from rat striatum slices.
  • Reduces dopamine levels in the striatum or limbic system at high doses.
  • Increases dopamine synthesis in rat striatum and limbic system.
  • Increases extracellular dopamine levels.
  • Decreases striatal ACh levels.
  • Increases the duration of swimming behavior in acute studies.