Learn More
Medchemexpress LLC Amisulpride hydrochloride | 81342-13-4 | MFCD00958763 | 99.8% | C17H28ClN3O4S | 100 MG
SDP
Catalog No. 5000354452
Encompass_Preferred
Amisulpride hydrochloride is a dopamine D2/D3 receptor antagonist, characterized by Kis of 2.8 and 3.2 nM for human dopamine D2 and D3 receptors, respectively. This compound is widely utilized in pharmacological research to investigate dopamine receptor activity and its associated physiological effects.
  • Dopamine D2/D3 receptor antagonist
  • Kis of 2.8 and 3.2 nM for human dopamine D2 and D3
  • Inhibits quinpirole-elicited [3H]thymidine incorporation
  • Increases [3H]dopamine release from rat striatum slices
  • Used in studies related to neurological and neurodegenerative diseases
Catalog No. 50-003-54452 Supplier Medchemexpress LLC Supplier No. HY14545A100MG
Amisulpride hydrochloride is a dopamine D2/D3 receptor antagonist, characterized by Kis of 2.8 and 3.2 nM for human dopamine D2 and D3 receptors, respectively. This compound is widely utilized in pharmacological research to investigate dopamine receptor activity and its associated physiological effects.
  • Dopamine D2/D3 receptor antagonist
  • Kis of 2.8 and 3.2 nM for human dopamine D2 and D3
  • Inhibits quinpirole-elicited [3H]thymidine incorporation
  • Increases [3H]dopamine release from rat striatum slices
  • Used in studies related to neurological and neurodegenerative diseases