Learn More
Apexbio Technology LLC Amitraz 33089-61-1 200mg
Supplier: Apexbio Technology LLC C6542200
Amitraz (CAS 33089-61-1) is a small-molecule agonist targeting alpha-adrenergic and octopamine receptors It is designed to activate these receptors thereby affecting neurotransmitter regulation within the insect central nervous system Amitraz exerts its biological activity primarily through receptor agonism impairing monoamine oxidase and prostaglandin biosynthesis pathways In veterinary research particularly in canine models amitraz demonstrates non-systemic acaricidal and insecticidal activity Reported IC50 values vary by insect species and experimental conditions typically within micromolar concentrations Based on these pharmacological properties amitraz holds research potential in the control of mite and tick infestations in veterinary settings