Learn More
Medchemexpress LLC Amitraz | 33089-61-1 | 99.7% | 250 MG
SDP
Catalog No. 5000414484
Encompass_Preferred
Amitraz is a non-systemic acaricide and insecticide that acts as an alpha-adrenergic agonist, interacting with octopamine receptors in the central nervous system. It also inhibits monoamine oxidase and prostaglandin synthesis, making it suitable for various research applications.
  • Non-systemic acaricide and insecticide
  • Acts as an alpha-adrenergic agonist
  • Interacts with octopamine receptors in the central nervous system
  • Inhibits monoamine oxidase
  • Inhibits prostaglandin synthesis
  • High purity of 99.65%
  • Solid appearance
  • White to light yellow color
Catalog No. 50-004-14484 Supplier Medchemexpress LLC Supplier No. 250MGHYB1111
Amitraz is a non-systemic acaricide and insecticide that acts as an alpha-adrenergic agonist, interacting with octopamine receptors in the central nervous system. It also inhibits monoamine oxidase and prostaglandin synthesis, making it suitable for various research applications.
  • Non-systemic acaricide and insecticide
  • Acts as an alpha-adrenergic agonist
  • Interacts with octopamine receptors in the central nervous system
  • Inhibits monoamine oxidase
  • Inhibits prostaglandin synthesis
  • High purity of 99.65%
  • Solid appearance
  • White to light yellow color