Learn More
Medchemexpress LLC Amitraz-d3 | 33089-61-1 | 99.5% | 1 MG
SDP
Catalog No. 5000413486
Encompass_Preferred
Amitraz-d3 is a deuterium-labeled version of Amitraz, a non-systemic acaricide and insecticide. Amitraz functions as an alpha-adrenergic agonist, interacts with octopamine receptors in the central nervous system, and inhibits monoamine oxidases and prostaglandin synthesis. This compound is intended for research purposes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13486 Supplier Medchemexpress LLC Supplier No. HYB1111S11MG
Amitraz-d3 is a deuterium-labeled version of Amitraz, a non-systemic acaricide and insecticide. Amitraz functions as an alpha-adrenergic agonist, interacts with octopamine receptors in the central nervous system, and inhibits monoamine oxidases and prostaglandin synthesis. This compound is intended for research purposes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.