Learn More
Medchemexpress LLC Amitriptyline-d3 hydrochloride | 342611-00-1 | MFCD01073983 | 99.5% | 316.88 g/mol | C20H21D3ClN | 5 MG
SDP
Catalog No. 5000221909
Encompass_Preferred
Amitriptyline-d3 hydrochloride is a deuterium-labeled analytical standard of the tricyclic antidepressant amitriptyline, provided as the hydrochloride salt for use as an internal standard, tracer, or reference material in LC-MS/MS and other quantitative analytical assays. It is intended for method development, validation, and accurate quantitation where isotopic labeling improves specificity and precision.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity suitable for quantitative analysis (99.5%).
  • Supplied as a hydrochloride salt for stability and solubility.
  • Molecular weight approximately 316.88 g/mol.
  • Pack sizes compatible with analytical workflows and trace-level applications.
Catalog No. 50-002-21909 Supplier Medchemexpress LLC Supplier No. HY1350965MG
Amitriptyline-d3 hydrochloride is a deuterium-labeled analytical standard of the tricyclic antidepressant amitriptyline, provided as the hydrochloride salt for use as an internal standard, tracer, or reference material in LC-MS/MS and other quantitative analytical assays. It is intended for method development, validation, and accurate quantitation where isotopic labeling improves specificity and precision.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity suitable for quantitative analysis (99.5%).
  • Supplied as a hydrochloride salt for stability and solubility.
  • Molecular weight approximately 316.88 g/mol.
  • Pack sizes compatible with analytical workflows and trace-level applications.