Learn More
Medchemexpress LLC Amitriptyline-d3 (hydrochloride) | 342611-00-1 | MFCD01073983 | 99.5% | C20H21D3ClN | 10MG
SDP
Catalog No. 5000259281
Encompass_Preferred
Amitriptyline-d3 (hydrochloride) is the deuterium-labeled hydrochloride salt of amitriptyline supplied as a high-purity analytical standard for use as a tracer and internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS.
  • Deuterium-labeled internal standard for quantitative analysis
  • High chemical purity (99.5%) and isotopic labeling consistency
  • Compatible with NMR, GC-MS, and LC-MS workflows
  • Supplied as a hydrochloride salt for stable handling
  • Available in small reference sizes suitable for method development
Catalog No. 50-002-59281 Supplier Medchemexpress LLC Supplier No. HY13509610MG
Amitriptyline-d3 (hydrochloride) is the deuterium-labeled hydrochloride salt of amitriptyline supplied as a high-purity analytical standard for use as a tracer and internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS.
  • Deuterium-labeled internal standard for quantitative analysis
  • High chemical purity (99.5%) and isotopic labeling consistency
  • Compatible with NMR, GC-MS, and LC-MS workflows
  • Supplied as a hydrochloride salt for stable handling
  • Available in small reference sizes suitable for method development