Learn More
Medchemexpress LLC Amitriptyline-d6 hydrochloride | 203645-63-0 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0527AS1MG
Encompass_Preferred
Amitriptyline-d6 hydrochloride is the deuterium-labeled version of Amitriptyline hydrochloride, an orally active tricyclic antidepressant (TCA). It primarily works by blocking SERT (Ki = 3.45 nM) and NET (Ki = 13.3 nM), which increases the concentrations of 5-hydroxytryptamine (5-HT) and norepinephrine (NE) in the synaptic cleft. It also acts as an agonist at α2A and TrkA/TrkB receptors, contributing to analgesic and neurotrophic effects by inhibiting cell apoptosis.
  • Increases concentrations of 5-hydroxytryptamine and norepinephrine in synaptic cleft
  • Contributes to analgesic and neurotrophic effects
  • Reduces inflammation, angiogenesis, and fibrosis
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-06886