Learn More
Lkt Laboratories Amitriptyline Hydrochloride, ≥98%, 10GR
SDP
Supplier:  Lkt Laboratories A523510G
Encompass_Preferred
FIASMA, σ1, RyR2, TrkA/B agonist; SERT, NET inhibitor; K+, Ca2+ channel blocker. CAS: 549-18-8. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4867