Learn More
Medchemexpress LLC Amitriptyline hydrochloride | 549-18-8 | 99.9% | 313.86 g/mol | C20H24ClN | 10 MG
SDP
Catalog No. 5000298066
Encompass_Preferred
Amitriptyline hydrochloride is an orally active tricyclic antidepressant compound provided as a laboratory reference standard for analytical and pharmacological studies. It is supplied as a solid with defined purity and solubility suitable for in vitro assays and method development.
  • High purity suitable for analytical use.
  • White to off-white solid form for easy handling.
  • Defined molecular weight and CAS number for accurate identification.
  • Stable when stored sealed at 4°C; solvent storage guidelines available.
  • Soluble in DMSO and water at stated concentrations for assay preparation.
  • Available in a 10 mg packaged quantity for small-scale studies.
Catalog No. 50-002-98066 Supplier Medchemexpress LLC Supplier No. HYB0527AR10MG
Amitriptyline hydrochloride is an orally active tricyclic antidepressant compound provided as a laboratory reference standard for analytical and pharmacological studies. It is supplied as a solid with defined purity and solubility suitable for in vitro assays and method development.
  • High purity suitable for analytical use.
  • White to off-white solid form for easy handling.
  • Defined molecular weight and CAS number for accurate identification.
  • Stable when stored sealed at 4°C; solvent storage guidelines available.
  • Soluble in DMSO and water at stated concentrations for assay preparation.
  • Available in a 10 mg packaged quantity for small-scale studies.