Learn More
Medchemexpress LLC Amitriptyline hydrochloride | 549-18-8 | 99.9% | 5 G
SDP
Catalog No. 5000407574
Encompass_Preferred
Amitriptyline hydrochloride is an orally active tricyclic antidepressant (TCA). It primarily exerts its antidepressant effect by blocking SERT and NET, which increases the concentrations of 5-hydroxytryptamine (5-HT) and norepinephrine (NE) in the synaptic cleft. It also acts as an agonist at α2A and TrkA/TrkB receptors, providing analgesic and neurotrophic activities. Amitriptyline hydrochloride can reduce inflammation, angiogenesis, and fibrosis.
  • Orally active tricyclic antidepressant (TCA)
  • Blocks SERT and NET, increasing 5-HT and NE concentrations
  • Agonist at α2A and TrkA/TrkB receptors
  • Provides analgesic and neurotrophic activities
  • Can reduce inflammation, angiogenesis, and fibrosis
  • Binds to DAT and antagonizes muscarinic, H1, and adrenergic α1 receptors
  • Can block sodium channels and hERG potassium channel
Catalog No. 50-004-07574 Supplier Medchemexpress LLC Supplier No. HYB0527A5G
Amitriptyline hydrochloride is an orally active tricyclic antidepressant (TCA). It primarily exerts its antidepressant effect by blocking SERT and NET, which increases the concentrations of 5-hydroxytryptamine (5-HT) and norepinephrine (NE) in the synaptic cleft. It also acts as an agonist at α2A and TrkA/TrkB receptors, providing analgesic and neurotrophic activities. Amitriptyline hydrochloride can reduce inflammation, angiogenesis, and fibrosis.
  • Orally active tricyclic antidepressant (TCA)
  • Blocks SERT and NET, increasing 5-HT and NE concentrations
  • Agonist at α2A and TrkA/TrkB receptors
  • Provides analgesic and neurotrophic activities
  • Can reduce inflammation, angiogenesis, and fibrosis
  • Binds to DAT and antagonizes muscarinic, H1, and adrenergic α1 receptors
  • Can block sodium channels and hERG potassium channel