Learn More
Sigma Aldrich Fine Chemicals Biosciences Amitriptyline N-AY-D-glucu
Catalog No. 502897094
Encompass
A certified SOLUTIONon standard of Amitriptyline N-AY-D-glucuronide a metabolite of the tricyclic antidepressant amitriptyline. Sold under trade names such as Amitrip Elavil and Levate amitriptyline is prescribed to treat a number of mental disorders ranging from major depressive disorder and anxiety disorder to bipolar disorder. This SOLUTIONon-based Certified Reference Material is applicable for use in LC/MS or GC/MS testing applications in clinical toxicology forensic analysis workplace drug testing and pharmaceutical research.
Catalog No. 50-289-7094 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A1281ML
A certified SOLUTIONon standard of Amitriptyline N-AY-D-glucuronide a metabolite of the tricyclic antidepressant amitriptyline. Sold under trade names such as Amitrip Elavil and Levate amitriptyline is prescribed to treat a number of mental disorders ranging from major depressive disorder and anxiety disorder to bipolar disorder. This SOLUTIONon-based Certified Reference Material is applicable for use in LC/MS or GC/MS testing applications in clinical toxicology forensic analysis workplace drug testing and pharmaceutical research.