Learn More
Medchemexpress LLC Amitriptyline-N-glucuronide-d3 | 112806-33-4 | 97.0% | C26H28D3NO6 | 1 MG
SDP
Catalog No. 5000355559
Encompass_Preferred
Amitriptyline-N-glucuronide-d3 is the deuterium labeled Amitriptyline-N-glucuronide. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, is used largely as a tracer for quantitation during drug development and has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-55559 Supplier Medchemexpress LLC Supplier No. HY143915S1MG
Amitriptyline-N-glucuronide-d3 is the deuterium labeled Amitriptyline-N-glucuronide. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, is used largely as a tracer for quantitation during drug development and has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS