Learn More
Cayman Chemical AMK (hydrochloride), Purity: ≥95%, CAS: 1215711-91-3, 10 mg
SDP
Catalog No. 502910739
Encompass_Preferred
An active metabolite of melatonin; scavenges singlet oxygen in vitro at 200 µM; inhibits epinephrine- and arachidonic acid-induced production of PGE2 and PGD2 in ovine seminal vesicle; inhibits LPS-induced increases in COX-2 levels in RAW 264.7 macrophages at 500 µM; decreases MPTP-induced increases in lipid peroxidation in the cytosol and mitochondria from substantia nigra and striatum in a mouse model of MPTP-induced Parkinson's disease at 20 mg/kg
Catalog No. 50-291-0739 Supplier Cayman Chemical Supplier No. 3468810 MG
An active metabolite of melatonin; scavenges singlet oxygen in vitro at 200 µM; inhibits epinephrine- and arachidonic acid-induced production of PGE2 and PGD2 in ovine seminal vesicle; inhibits LPS-induced increases in COX-2 levels in RAW 264.7 macrophages at 500 µM; decreases MPTP-induced increases in lipid peroxidation in the cytosol and mitochondria from substantia nigra and striatum in a mouse model of MPTP-induced Parkinson's disease at 20 mg/kg