Learn More
ATCC AML12; Hepatocyte; Mouse (Mus musculus)
Catalog No. 502384510
Encompass
AML12 (alpha mouse liver 12) cells are hepatocytes isolated from the normal liver of a 3-month-old mouse. Use these cells as a transfection host or in toxicology research applications.
Catalog No. 50-238-4510 Supplier ATCC Supplier No. CRL2254
AML12 (alpha mouse liver 12) cells are hepatocytes isolated from the normal liver of a 3-month-old mouse. Use these cells as a transfection host or in toxicology research applications.