Learn More
Cayman Chemical Amlodipine-d4 (maleate), Purity: ≥99% deuterated forms (d1-d4), CAS: 1185246-15-4, 5 mg
SDP
Catalog No. 502934672
Encompass_Preferred
An internal standard for the quantification of amlodipine by GC- or LC-MS
Catalog No. 50-293-4672 Supplier Cayman Chemical Supplier No. 250355 MG
An internal standard for the quantification of amlodipine by GC- or LC-MS