Learn More
Cayman Chemical Amlodipine (besylate), Purity: ≥98%, CAS: 111470-99-6, 1 g
SDP
Catalog No. 502970041
Encompass_Preferred
An L-type calcium channel blocker; inhibits calcium influx in cardiac and vascular smooth muscle; inhibits calcium-induced contractions in depolarized rat aorta (IC50 = 1.9 nM), displaying a slow rate of association and dissociation in isolated vascular and cardiac tissues
Catalog No. 50-297-0041 Supplier Cayman Chemical Supplier No. 148861 G
An L-type calcium channel blocker; inhibits calcium influx in cardiac and vascular smooth muscle; inhibits calcium-induced contractions in depolarized rat aorta (IC50 = 1.9 nM), displaying a slow rate of association and dissociation in isolated vascular and cardiac tissues