Learn More
Medchemexpress LLC Amlodipine-1,1,2,2-d4 | 1185246-14-3 | MFCD21363414 | 98.5% | 412.90 | C20H21D4ClN2O5 | 10 MG
SDP
Catalog No. 5000298228
Encompass_Preferred
Amlodipine-1,1,2,2-d4 is a deuterium-labeled analog of amlodipine supplied as a solid analytical standard for research. It is intended for use as an internal standard or tracer in pharmacokinetic, bioanalytical, and isotopic-labeling studies involving L-type calcium channel blockers.
  • Deuterium-labeled (1,1,2,2-d4) analog suitable as an internal standard.
  • High purity suitable for analytical applications.
  • Molecular weight 412.90 for accurate mass-based analyses.
  • Supplied as a solid in small mg quantities for laboratory use.
  • Common applications include pharmacokinetic and bioanalytical studies.
  • Available in typical package sizes of 5 mg, 10 mg, and 50 mg.
Catalog No. 50-002-98228 Supplier Medchemexpress LLC Supplier No. HYB0317S10MG
Amlodipine-1,1,2,2-d4 is a deuterium-labeled analog of amlodipine supplied as a solid analytical standard for research. It is intended for use as an internal standard or tracer in pharmacokinetic, bioanalytical, and isotopic-labeling studies involving L-type calcium channel blockers.
  • Deuterium-labeled (1,1,2,2-d4) analog suitable as an internal standard.
  • High purity suitable for analytical applications.
  • Molecular weight 412.90 for accurate mass-based analyses.
  • Supplied as a solid in small mg quantities for laboratory use.
  • Common applications include pharmacokinetic and bioanalytical studies.
  • Available in typical package sizes of 5 mg, 10 mg, and 50 mg.