Learn More
Medchemexpress LLC Amlodipine-1,1,2,2-d4 maleate | 1185246-15-4 | 98.0% | 5 MG
SDP
Catalog No. 5000406674
Encompass_Preferred
Amlodipine-1,1,2,2-d4 maleate is the deuterium-labeled form of Amlodipine, an antianginal agent and an orally active dihydropyridine calcium channel blocker. It works by blocking voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium. This compound is intended for research use only.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06674 Supplier Medchemexpress LLC Supplier No. HYB0317S5MG
Amlodipine-1,1,2,2-d4 maleate is the deuterium-labeled form of Amlodipine, an antianginal agent and an orally active dihydropyridine calcium channel blocker. It works by blocking voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium. This compound is intended for research use only.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.