Learn More
Medchemexpress LLC Amlodipine-1,1,2,2-d4 maleate | 1185246-15-4 | 98.04% | 1 MG
SDP
Catalog No. 5000406808
Encompass_Preferred
Amlodipine-1,1,2,2-d4 maleate is a deuterium-labeled calcium channel blocker that acts as an antianginal agent. It inhibits calcium influx by blocking voltage-dependent L-type calcium channels. This compound is utilized in research for high blood pressure and cancer, and can serve as a tracer or internal standard for quantitative analysis via NMR, GC-MS, or LC-MS. Deuterium labeling provides a means for precise quantitation in drug development and can influence drug pharmacokinetics and metabolism.
  • Deuterium labeled calcium channel blocker
  • Acts as an antianginal agent
  • Inhibits calcium influx by blocking L-type channels
  • Used in research for high blood pressure and cancer
  • Functions as a tracer or internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, LC-MS
  • Deuteration influences drug pharmacokinetics and metabolism
Catalog No. 50-004-06808 Supplier Medchemexpress LLC Supplier No. HYB0317S1MG
Amlodipine-1,1,2,2-d4 maleate is a deuterium-labeled calcium channel blocker that acts as an antianginal agent. It inhibits calcium influx by blocking voltage-dependent L-type calcium channels. This compound is utilized in research for high blood pressure and cancer, and can serve as a tracer or internal standard for quantitative analysis via NMR, GC-MS, or LC-MS. Deuterium labeling provides a means for precise quantitation in drug development and can influence drug pharmacokinetics and metabolism.
  • Deuterium labeled calcium channel blocker
  • Acts as an antianginal agent
  • Inhibits calcium influx by blocking L-type channels
  • Used in research for high blood pressure and cancer
  • Functions as a tracer or internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, LC-MS
  • Deuteration influences drug pharmacokinetics and metabolism