Learn More
Enzo Life Sciences Amlodipine (1g). CAS: 88150-42-9
SDP
Supplier:  Enzo Life Sciences BMLCA2021000
Encompass_Preferred
Amlodipine is a third generation L-type calcium channel blocker with a unique spectrum of actions, over and above its antihypertensive activity which include stimulation of NO synthase, inhibition of monocyte-endothelial adhesion, inhibition of oxidative stress and sustained duration of action. Purity: ≥98% (Assay). Solubility: Soluble in methanol, DMSO (25mg/ml), 100% ethanol (25mg/ml), dimethyl formamide (25mg/ml) or chloroform. Long Term Storage: Ambient.
Catalog No. 50-201-0360