Learn More
Medchemexpress LLC HY-B0317 500mg Medchemexpress, Amlodipine CAS:88150-42-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0317500MG
Encompass_Preferred
Medchemexpress, HY-B0317 500mg Amlodipine CAS:88150-42-9 Amlodipine, an antianginal agent and an orally active dihydropyridine calcium channel blocker, works by blocking the voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2464