Learn More
Selleck Chemical LLC Amlodipine-50mg
SDP
Supplier:  Selleck Chemical LLC S190550MG
Encompass_Preferred
Amlodipine (UK-48340) is a long-acting calcium channel blocker, used to lower blood pressure and prevent chest pain.
Catalog No. 50-238-9583