Learn More
Apexbio Technology LLC Amlodipine 88150-42-9 1g
SDP
Catalog No. 5000089342
Encompass_Preferred
Amlodipine (CAS 88150-42-9) is a dihydropyridine-based small molecule that functions as a long-acting calcium channel blocker It selectively inhibits L-type calcium channels on vascular smooth muscle and cardiac myocytes thereby reducing intracellular calcium influx This action leads to vasodilation and decreased vascular resistance Amlodipine is widely utilized in biomedical research to investigate calcium signaling pathways vascular tone modulation and cardiovascular pharmacology Its mechanistic profile also makes it a reference compound in studies evaluating novel antihypertensive or vasodilatory agents
Catalog No. 50-000-89342 Supplier Apexbio Technology LLC Supplier No. B14101000
Amlodipine (CAS 88150-42-9) is a dihydropyridine-based small molecule that functions as a long-acting calcium channel blocker It selectively inhibits L-type calcium channels on vascular smooth muscle and cardiac myocytes thereby reducing intracellular calcium influx This action leads to vasodilation and decreased vascular resistance Amlodipine is widely utilized in biomedical research to investigate calcium signaling pathways vascular tone modulation and cardiovascular pharmacology Its mechanistic profile also makes it a reference compound in studies evaluating novel antihypertensive or vasodilatory agents