Learn More
Medchemexpress LLC Amlodipine | 88150-42-9 | 99.87% | 5 G
SDP
Catalog No. 5000406903
Encompass_Preferred
Amlodipine is an antianginal agent and an orally active dihydropyridine calcium channel blocker. It functions by blocking voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium. It can be used in the research of high blood pressure and cancer.
  • Functions as an antianginal agent
  • Orally active dihydropyridine calcium channel blocker
  • Blocks voltage-dependent L-type calcium channels
  • Inhibits initial influx of calcium
  • Suitable for research on high blood pressure
  • Suitable for research on cancer
Catalog No. 50-004-06903 Supplier Medchemexpress LLC Supplier No. HYB03175G
Amlodipine is an antianginal agent and an orally active dihydropyridine calcium channel blocker. It functions by blocking voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium. It can be used in the research of high blood pressure and cancer.
  • Functions as an antianginal agent
  • Orally active dihydropyridine calcium channel blocker
  • Blocks voltage-dependent L-type calcium channels
  • Inhibits initial influx of calcium
  • Suitable for research on high blood pressure
  • Suitable for research on cancer