Learn More
Medchemexpress LLC Amlodipine | 88150-42-9 | MFCD00864687 | 99.9% | 1 ML
SDP
Catalog No. 5000406775
Encompass_Preferred
Amlodipine is an antianginal agent and an orally active dihydropyridine calcium channel blocker. It functions by blocking voltage-dependent L-type calcium channels, inhibiting the initial influx of calcium. This compound is used in research for its effects on high blood pressure and cancer.
  • Orally active dihydropyridine calcium channel blocker
  • Blocks voltage-dependent L-type calcium channels
  • Utilized in research for high blood pressure
  • Utilized in research for cancer
Catalog No. 50-004-06775 Supplier Medchemexpress LLC Supplier No. HYB031710MM/1ML
Amlodipine is an antianginal agent and an orally active dihydropyridine calcium channel blocker. It functions by blocking voltage-dependent L-type calcium channels, inhibiting the initial influx of calcium. This compound is used in research for its effects on high blood pressure and cancer.
  • Orally active dihydropyridine calcium channel blocker
  • Blocks voltage-dependent L-type calcium channels
  • Utilized in research for high blood pressure
  • Utilized in research for cancer