Learn More
Medchemexpress LLC Amlodipine benzenesulfonate | 111470-99-6 | 99.7% | 500 MG
SDP
Catalog No. 5000406822
Encompass_Preferred
Amlodipine besylate (Amlodipine benzenesulfonate) is an orally active dihydropyridine calcium channel blocker and antianginal agent. It functions by blocking voltage-dependent L-type calcium channels, inhibiting initial calcium influx. This compound can be used for research into high blood pressure and cancer.
  • Blocks voltage-dependent L-type calcium channels
  • Inhibits initial influx of calcium
  • Useful for high blood pressure research
  • Useful for cancer research
Catalog No. 50-004-06822 Supplier Medchemexpress LLC Supplier No. HYB0317B500MG
Amlodipine besylate (Amlodipine benzenesulfonate) is an orally active dihydropyridine calcium channel blocker and antianginal agent. It functions by blocking voltage-dependent L-type calcium channels, inhibiting initial calcium influx. This compound can be used for research into high blood pressure and cancer.
  • Blocks voltage-dependent L-type calcium channels
  • Inhibits initial influx of calcium
  • Useful for high blood pressure research
  • Useful for cancer research