Learn More
Medchemexpress LLC Amlodipine besylate | 111470-99-6 | 99.7% | 1 ML
SDP
Catalog No. 5000406919
Encompass_Preferred
Amlodipine besylate is an antianginal agent and an orally active dihydropyridine calcium channel blocker. It functions by blocking voltage-dependent L-type calcium channels, which inhibits the initial influx of calcium. This product is for research use only and not sold to patients.
  • Used for high blood pressure and cancer research
  • Inhibits initial influx of calcium by blocking L-type channels
  • Available as 10 mM in 1 mL solution in DMSO
Catalog No. 50-004-06919 Supplier Medchemexpress LLC Supplier No. HYB0317B10MM/1ML
Amlodipine besylate is an antianginal agent and an orally active dihydropyridine calcium channel blocker. It functions by blocking voltage-dependent L-type calcium channels, which inhibits the initial influx of calcium. This product is for research use only and not sold to patients.
  • Used for high blood pressure and cancer research
  • Inhibits initial influx of calcium by blocking L-type channels
  • Available as 10 mM in 1 mL solution in DMSO