Learn More
Apexbio Technology LLC Amlodipine Besylate 111470-99-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062198
Encompass_Preferred
Amlodipine Besylate (CAS 111470-99-6) is a long-acting calcium channel blocker that reduces blood pressure primarily through inhibition of calcium ion influx via voltage-gated calcium channels Calcium channel modulation influences critical cellular signaling pathways In cortical neurons exposed to oxidative stress Amlodipine Besylate ( 5 M) preserves cellular viability by reducing oxidative damage enhancing pro-survival signals and suppressing apoptotic pathways It has also demonstrated proliferative effects on human epidermoid carcinoma (A431) cells Furthermore clinical studies indicate that Amlodipine Besylate decreases peripheral vascular resistance and myocardial oxygen demand in angina patients and effectively lowers arterial pressure in hypertensive feline models
Catalog No. 50-000-62198 Supplier Apexbio Technology LLC Supplier No. B14115.1
Amlodipine Besylate (CAS 111470-99-6) is a long-acting calcium channel blocker that reduces blood pressure primarily through inhibition of calcium ion influx via voltage-gated calcium channels Calcium channel modulation influences critical cellular signaling pathways In cortical neurons exposed to oxidative stress Amlodipine Besylate ( 5 M) preserves cellular viability by reducing oxidative damage enhancing pro-survival signals and suppressing apoptotic pathways It has also demonstrated proliferative effects on human epidermoid carcinoma (A431) cells Furthermore clinical studies indicate that Amlodipine Besylate decreases peripheral vascular resistance and myocardial oxygen demand in angina patients and effectively lowers arterial pressure in hypertensive feline models