Learn More
Selleck Chemical LLC Amlodipine Besylate S1813-1g
SDP
Catalog No. 503110802
Encompass_Preferred
Amlodipine Besylate is a long-acting calcium channel blocker used to lower blood pressure and prevent chest pain
Catalog No. 50-311-0802 Supplier Selleck Chemical LLC Supplier No. S18131G
Amlodipine Besylate is a long-acting calcium channel blocker used to lower blood pressure and prevent chest pain